Please login to your online account to display your discounted pricing

Methyle3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
CAS 175201-72-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H13NO4S2
Formula Weight 263.34
MDL Number MFCD00067930
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM01952DA

  SDS
fisher scientific company
KM01952DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM01952EA

  SDS
fisher scientific company
KM01952EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H13NO4S2
MDL Number MFCD00067930
CAS 175201-72-6