Please login to your online account to display your discounted pricing

Methyle3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM01952DA

 View more versions of this product

Catalog No. KM01952DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 263.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C9H13NO4S2
MDL Number MFCD00067930
CAS 175201-72-6

Specifications

Chemical Name or Material Methyl 3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
CAS 175201-72-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H13NO4S2
Formula Weight 263.34
MDL Number MFCD00067930