Please login to your online account to display your discounted pricing

Methyle3-amino-2-pyrazinecarboxylate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 3-amino-2-pyrazinecarboxylate
Name Note 98%
CAS 16298-03-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7N3O2
Formula Weight 153.14
MDL Number MFCD00010097
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269590100

  SDS
fisher scientific acros
AC269590100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 153.14
Quantity 10g
Color Yellow
Beilstein 25, IV, 4412
Melting Point 166° to 172°C
Infrared Spectrum Authentic
MDL Number MFCD00010097
Assay Percent Range 98%
Chemical Name or Material Methyl 3-amino-2-pyrazinecarboxylate
Packaging Glass bottle
Molecular Formula C6H7N3O2
Name Note 98%
CAS 16298-03-6
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.