Please login to your online account to display your discounted pricing

Methyle3-amino-2-pyrazinecarboxylate, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC269590100

 View more versions of this product

Catalog No. AC269590100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 153.14
Quantity 10g
Color Yellow
Beilstein 25, IV, 4412
Melting Point 166° to 172°C
Chemical Name or Material Methyl 3-amino-2-pyrazinecarboxylate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H7N3O2
MDL Number MFCD00010097
Infrared Spectrum Authentic
Name Note 98%
CAS 16298-03-6
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl 3-amino-2-pyrazinecarboxylate
Name Note 98%
CAS 16298-03-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7N3O2
Formula Weight 153.14
MDL Number MFCD00010097
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents