Please login to your online account to display your discounted pricing

Methyle3-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate
CAS 175203-23-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15BrN2O2
Formula Weight 323.19
MDL Number MFCD00068047
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM06349EA

  SDS
fisher scientific company
KM06349EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM06349DA

  SDS
fisher scientific company
KM06349DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 323.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C14H15BrN2O2
CAS 175203-23-3
MDL Number MFCD00068047
Chemical Name or Material Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate
Assay Percent Range 97%