Please login to your online account to display your discounted pricing

Methyle3-[(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM06349EA

 View more versions of this product

Catalog No. KM06349EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 323.19
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C14H15BrN2O2
MDL Number MFCD00068047
CAS 175203-23-3
Chemical Name or Material Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate
Assay Percent Range 97%

Specifications

Chemical Name or Material Methyl 3-[(4-bromo-3, 5-dimethyl-1H-pyrazol-1-yl)methyl]benzoate
CAS 175203-23-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15BrN2O2
Formula Weight 323.19
MDL Number MFCD00068047