missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97%, ACROS Organics™

Chemical Identifiers

CAS 480425-35-2
Assay Percent Range 97%
Molecular Formula C14H19BO4
Formula Weight 262.11
MDL Number MFCD03789258
Chemical Name or Material Methyle3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC367550050 SDS Thermo Fisher Scientific
AC367550050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

480425-35-2
C14H19BO4
MFCD03789258
≥96.0%
97%
262.11
Methyle3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97%

Specifications

White
5g
Authentic
97%
262.11
Methyle3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 97%
90° to 97°C
480425-35-2
Crystalline Powder
C14H19BO4
MFCD03789258
≥96.0%

Safety and Handling

Recommended Storage : Refrigerator +4°C