Please login to your online account to display your discounted pricing

Methyle2-methyl-1,3-benzoxazole-5-carboxylate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle2-methyl-1, 3-benzoxazole-5-carboxylate
CAS 136663-21-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H9NO3
Formula Weight 191.19
MDL Number MFCD07774245
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432400050

  SDS
thermo fisher scientific
AC432400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432400010

  SDS
thermo fisher scientific
AC432400010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 191.19
Quantity 5g
Color White
Melting Point 65° to 69°C
Chemical Name or Material Methyle2-methyl-1, 3-benzoxazole-5-carboxylate
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H9NO3
MDL Number MFCD07774245
CAS 136663-21-3
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.