Please login to your online account to display your discounted pricing

Methyle2-isothiocyanatobenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2-isothiocyanatobenzoate
CAS 16024-82-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7NO2S
Linear Formula SCNC6H4CO2CH3
Formula Weight 193.23
MDL Number MFCD00041062
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01695EA

 
fisher scientific company
SB01695EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.23
Quantity 10g
Assay 97%
Density 1.2500g/mL
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula SCNC6H4CO2CH3
MDL Number MFCD00041062
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber glass bottle
Chemical Name or Material Methyl 2-isothiocyanatobenzoate
Molecular Formula C9H7NO2S
CAS 16024-82-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.