missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle2-cyclopentanonecarboxylate, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC342091000

 View more versions of this product

Catalog No. AC342091000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Clear colorless to slight yellow
142.15
100g
10, 597
C7H10O3
Methyle2-cyclopentanonecarboxylate
1.4550 to 1.4570 (20°C, 589nm)
Authentic
Liquid
≥95%
1.054g/mL
96%
10472-24-9
MFCD00001411
Glass bottle

Chemical Identifiers

10472-24-9
C7H10O3
MFCD00001411
≥95%
96%
142.15
Methyle2-cyclopentanonecarboxylate