Please login to your online account to display your discounted pricing

Methyle2-(chloromethyl)acrylate, 97%, stabilised with 0.1% hydroquinone, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 2-(chloromethyl)acrylate
Name Note Stabilized
CAS 922-15-6
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C5H7ClO2
Formula Weight 134.56g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC436110050

 
thermo fisher scientific
436110050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Stabilizer 0.1% (hydroquinone)
Color Clear colorless to pale yellow
Quantity 5g
Chemical Name or Material Methyl 2-(chloromethyl)acrylate
Name Note Stabilized
CAS 922-15-6
Physical Form Clear colorless to pale yellow liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C5H7ClO2
Formula Weight 134.56g/mol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.