Please login to your online account to display your discounted pricing

Methyle2-(chloromethyl)acrylate, 97%, stabilised with 0.1% hydroquinone, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC436110050

 View more versions of this product

Catalog No. AC436110050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 134.56
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
Chemical Name or Material Methyl 2-(chloromethyl)acrylate
Name Note Stabilized
Assay Percent Range 97%
CAS 922-15-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Stabilizer 0.1% (hydroquinone)
Molecular Formula C5H7ClO2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl 2-(chloromethyl)acrylate
Name Note Stabilized
CAS 922-15-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H7ClO2
Formula Weight 134.56
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.