Please login to your online account to display your discounted pricing

Methyle2-amino-4-chlorobenzoate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 2-amino-4-chlorobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8ClNO2
Formula Weight 185.61
Synonym Methyl 4-chloroanthranilate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC439231000

 
thermo fisher scientific
AC439231000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥97.5%
Formula Weight 185.61
Quantity 100g
Color White to pale brown
Melting Point 65° to 71°C
Packaging Glass bottle
Synonym Methyl 4-chloroanthranilate
Molecular Formula C8H8ClNO2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 2-amino-4-chlorobenzoate
Assay Percent Range 98%
CAS 5900-58-3
Health Hazard 1 Warning
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.