Please login to your online account to display your discounted pricing

Methyle2,6-dichloroisonicotinate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2, 6-dichloroisonicotinate
CAS 42521-09-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5Cl2NO2
Formula Weight 206.03
MDL Number MFCD00044409
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB05937DA

  SDS
fisher scientific company
SPB05937DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.03
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 42521-09-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00044409
Molecular Formula C7H5Cl2NO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 2, 6-dichloroisonicotinate
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.