missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Methyle2,4-dichlorophenylacetate, 99%, ACROS Organics™
Chemical Identifiers
CAS
55954-23-9
Molecular Formula
C9H8Cl2O2
Molecular Weight (g/mol)
219.061
MDL Number
MFCD00018381
InChI Key
SRCVNBFTEARRJY-UHFFFAOYSA-N
Synonym
2,4-dichlorobenzeneacetic acid methyl ester, 2,4-dichlorophenylacetic acid methyl ester, 2,4-dichlorophenylaceticacidmethylester, acmc-209lq7, benzeneacetic acid, 2,4-dichloro-, methyl ester, dcaa methyl ester, methyl 2,4-dichlorobenzeneacetate, methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, methyl2,4-dichlorophenylacetate
PubChem CID
2734106
IUPAC Name
methyl 2-(2,4-dichlorophenyl)acetate
SMILES
COC(=O)CC1=C(C=C(C=C1)Cl)Cl
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
AC351060250
SDS
View Documents
Thermo Fisher Scientific
351060250
25g
Glass Bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
55954-23-9
Molecular Weight (g/mol)
219.061
InChI Key
SRCVNBFTEARRJY-UHFFFAOYSA-N
PubChem CID
2734106
SMILES
COC(=O)CC1=C(C=C(C=C1)Cl)Cl
Molecular Formula
C9H8Cl2O2
MDL Number
MFCD00018381
Synonym
2,4-dichlorobenzeneacetic acid methyl ester, 2,4-dichlorophenylacetic acid methyl ester, 2,4-dichlorophenylaceticacidmethylester, acmc-209lq7, benzeneacetic acid, 2,4-dichloro-, methyl ester, dcaa methyl ester, methyl 2,4-dichlorobenzeneacetate, methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, methyl2,4-dichlorophenylacetate
IUPAC Name
methyl 2-(2,4-dichlorophenyl)acetate
Specifications
Infrared Spectrum
Authentic
Refractive Index
1.5390 to 1.5420
Quantity
25g
Assay Percent Range
99%
MDL Number
MFCD00018381
InChI Key
SRCVNBFTEARRJY-UHFFFAOYSA-N
IUPAC Name
methyl 2-(2,4-dichlorophenyl)acetate
PubChem CID
2734106
Physical Form
Liquid
Chemical Name or Material
Methyl 2,4-dichlorophenylacetate
Packaging
Glass Bottle
Color
Colorless to Yellow
CAS
55954-23-9
Molecular Formula
C9H8Cl2O2
Synonym
2,4-dichlorobenzeneacetic acid methyl ester, 2,4-dichlorophenylacetic acid methyl ester, 2,4-dichlorophenylaceticacidmethylester, acmc-209lq7, benzeneacetic acid, 2,4-dichloro-, methyl ester, dcaa methyl ester, methyl 2,4-dichlorobenzeneacetate, methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, methyl2,4-dichlorophenylacetate
SMILES
COC(=O)CC1=C(C=C(C=C1)Cl)Cl
Molecular Weight (g/mol)
219.061
Formula Weight
219.07
Percent Purity
99%
Safety and Handling
Safety and Handling
GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS