Learn More
Methyle2,4-dichlorophenylacetate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 351060250
Specifications
Methyl 2,4-dichlorophenylacetate | |
Glass Bottle | |
Colorless to Yellow | |
55954-23-9 | |
C9H8Cl2O2 | |
2,4-dichlorobenzeneacetic acid methyl ester, 2,4-dichlorophenylacetic acid methyl ester, 2,4-dichlorophenylaceticacidmethylester, acmc-209lq7, benzeneacetic acid, 2,4-dichloro-, methyl ester, dcaa methyl ester, methyl 2,4-dichlorobenzeneacetate, methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, methyl2,4-dichlorophenylacetate | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl | |
219.061 | |
219.07 | |
99% |
Authentic | |
1.5390 to 1.5420 | |
25g | |
99% | |
MFCD00018381 | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
methyl 2-(2,4-dichlorophenyl)acetate | |
2734106 | |
Liquid |
Chemical Identifiers
55954-23-9 | |
219.061 | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
2734106 | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl |
C9H8Cl2O2 | |
MFCD00018381 | |
2,4-dichlorobenzeneacetic acid methyl ester, 2,4-dichlorophenylacetic acid methyl ester, 2,4-dichlorophenylaceticacidmethylester, acmc-209lq7, benzeneacetic acid, 2,4-dichloro-, methyl ester, dcaa methyl ester, methyl 2,4-dichlorobenzeneacetate, methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, methyl2,4-dichlorophenylacetate | |
methyl 2-(2,4-dichlorophenyl)acetate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.