Please login to your online account to display your discounted pricing

Methyle2,4-dichlorobenzoate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle2, 4-dichlorobenzoate
CAS 35112-28-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6Cl2O2
Formula Weight 205.04
MDL Number MFCD00085117
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC430050050

  SDS
thermo fisher scientific
AC430050050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430050250

  SDS
thermo fisher scientific
AC430050250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 205.04
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00085117
Molecular Formula C8H6Cl2O2
Assay Percent Range 97%
Chemical Name or Material Methyle2, 4-dichlorobenzoate
CAS 35112-28-8
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.