missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle2-(3-methoxyphenyl)acetate, 97%, Maybridge

Chemical Identifiers

CAS 18927-05-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00017205
InChI Key BSVIOYCZTJRBDB-UHFFFAOYSA-N
Synonym 3-methoxy-phenyl-acetic acid methyl ester
PubChem CID 519609
IUPAC Name methyl 2-(3-methoxyphenyl)acetate
SMILES COC1=CC=CC(=C1)CC(=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
BTB09252DA SDS missing translation for 'viewDocuments' Fisher Scientific Company
BTB09252DA
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

18927-05-4
180.203
BSVIOYCZTJRBDB-UHFFFAOYSA-N
519609
COC1=CC=CC(=C1)CC(=O)OC
C10H12O3
MFCD00017205
3-methoxy-phenyl-acetic acid methyl ester
methyl 2-(3-methoxyphenyl)acetate

Specifications

Amber glass bottle
18927-05-4
100.0
C10H12O3
MFCD00017205
BSVIOYCZTJRBDB-UHFFFAOYSA-N
methyl 2-(3-methoxyphenyl)acetate
519609
97%
1g
96.5
97%
CH3OC6H4CH2C(O)OCH3
3-methoxy-phenyl-acetic acid methyl ester
COC1=CC=CC(=C1)CC(=O)OC
180.203
180.2
Methyle2-(3-methoxyphenyl)acetate