Please login to your online account to display your discounted pricing

Methyle2-(3-methoxyphenyl)acetate, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB09252DA

 View more versions of this product

Catalog No. BTB09252DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.2
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Methyle2-(3-methoxyphenyl)acetate
Linear Formula CH3OC6H4CH2C(O)OCH3
CAS Min % 96.5
MDL Number MFCD00017205
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C10H12O3
CAS 18927-05-4

Specifications

Chemical Name or Material Methyle2-(3-methoxyphenyl)acetate
CAS 18927-05-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2C(O)OCH3
Formula Weight 180.2
MDL Number MFCD00017205