Please login to your online account to display your discounted pricing

Methyle2,2,2-trichloroacetimidate, 98%, ACROS Organics™

$155.81 - $155.81

Chemical Identifiers

Chemical Name or Material Methyle2, 2, 2-trichloroacetimidate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H4Cl3NO
Linear Formula CCl3C(=NH)OCH3
Formula Weight 176.43
MDL Number MFCD00001759
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160931000

  SDS
thermo fisher scientific
AC160931000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160930250

  SDS
fisher scientific acros
160930250
Glass bottle 25g Each for $155.81
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 176.43
Quantity 100g
Color Clear colorless to pale yellow
Density 1.42g/mL
Beilstein 02, 212
Chemical Name or Material Methyle2, 2, 2-trichloroacetimidate
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CCl3C(=NH)OCH3
CAS 2533-69-9
Refractive Index 1.4770 to 1.4800 (20°C, 589nm)
Molecular Formula C3H4Cl3NO
MDL Number MFCD00001759
Packaging Glass bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.