Please login to your online account to display your discounted pricing

Methyle1H-benzimidazole-5-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 1H-benzimidazole-5-carboxylate
CAS 26663-77-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2O2
Formula Weight 176.17
MDL Number MFCD03407310
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00298CB

  SDS
fisher scientific company
MO00298CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.17
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 1H-benzimidazole-5-carboxylate
MDL Number MFCD03407310
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8N2O2
CAS 26663-77-4