Please login to your online account to display your discounted pricing

Methyle1H-benzimidazole-5-carboxylate, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00298CB

 View more versions of this product

Catalog No. MO00298CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.17
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 1H-benzimidazole-5-carboxylate
MDL Number MFCD03407310
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H8N2O2
CAS 26663-77-4

Specifications

Chemical Name or Material Methyl 1H-benzimidazole-5-carboxylate
CAS 26663-77-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2O2
Formula Weight 176.17
MDL Number MFCD03407310