missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436621000
Specifications
Methyl 1-aminocyclopropanecarboxylate hydrochloride | |
Authentic | |
176.4°C to 178.7°C | |
72784-42-0 | |
C5H9NO2·ClH | |
CHSDCWKLSHPKFY-UHFFFAOYSA-N | |
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride | |
12760695 | |
Crystalline Powder |
38.37 to 40.33% C, 8.95 to 9.40% | |
White | |
100mg | |
98% | |
1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropane-1-carboxylate hcl, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, methyl 1-aminocyclopropanecarboxylate hcl, methyl 1-aminocyclopropanecarboxylate hydrochloride, methyl-1-aminocyclopropanecarboxylate hydrochloride | |
COC(=O)C1(CC1)N.Cl | |
151.59 | |
151.59 | |
98% |
Chemical Identifiers
72784-42-0 | |
151.59 | |
1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropane-1-carboxylate hcl, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, methyl 1-aminocyclopropanecarboxylate hcl, methyl 1-aminocyclopropanecarboxylate hydrochloride, methyl-1-aminocyclopropanecarboxylate hydrochloride | |
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride |
C5H9NO2·ClH | |
CHSDCWKLSHPKFY-UHFFFAOYSA-N | |
12760695 | |
COC(=O)C1(CC1)N.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning