missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 72784-42-0
Molecular Formula C5H9NO2·ClH
Molecular Weight (g/mol) 151.59
InChI Key CHSDCWKLSHPKFY-UHFFFAOYSA-N
Synonym 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropane-1-carboxylate hcl, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, methyl 1-aminocyclopropanecarboxylate hcl, methyl 1-aminocyclopropanecarboxylate hydrochloride, methyl-1-aminocyclopropanecarboxylate hydrochloride
PubChem CID 12760695
IUPAC Name methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
SMILES COC(=O)C1(CC1)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC436621000
SDS View Documents
Thermo Fisher Scientific
AC436621000
100mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436625000
SDS View Documents
Thermo Fisher Scientific
AC436625000
500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

72784-42-0
151.59
1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropane-1-carboxylate hcl, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, methyl 1-aminocyclopropanecarboxylate hcl, methyl 1-aminocyclopropanecarboxylate hydrochloride, methyl-1-aminocyclopropanecarboxylate hydrochloride
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
C5H9NO2·ClH
CHSDCWKLSHPKFY-UHFFFAOYSA-N
12760695
COC(=O)C1(CC1)N.Cl

Specifications

38.37 to 40.33% C, 8.95 to 9.40%
White
100mg
98%
1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropane-1-carboxylate hcl, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, methyl 1-aminocyclopropanecarboxylate hcl, methyl 1-aminocyclopropanecarboxylate hydrochloride, methyl-1-aminocyclopropanecarboxylate hydrochloride
COC(=O)C1(CC1)N.Cl
151.59
151.59
98%
Authentic
176.4°C to 178.7°C
72784-42-0
C5H9NO2·ClH
CHSDCWKLSHPKFY-UHFFFAOYSA-N
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride
12760695
Crystalline Powder
Methyl 1-aminocyclopropanecarboxylate hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS