Please login to your online account to display your discounted pricing

Methyleα-aminoisobutyrate hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyleα-aminoisobutyrate hydrochloride
CAS 15028-41-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H11NO2·HCl
Formula Weight 151.64
MDL Number MFCD00214247
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC397980010

  SDS
thermo fisher scientific
AC397980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC397980050

  SDS
thermo fisher scientific
AC397980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 151.64
Quantity 1g
Color White
Assay Percent Range 98%
Molecular Formula C5H11NO2·HCl
CAS 15028-41-8
Chemical Name or Material Methyleα-aminoisobutyrate hydrochloride
NMR Consistent with structure
Packaging Glass bottle
MDL Number MFCD00214247
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.