Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyldiphenylphosphine, 99%

$36.03 - $527.97

Chemical Identifiers

Chemical Name or Material Methyldiphenylphosphine
CAS 1486-28-8
Molecular Formula (C6H5)2PCH3
Formula Weight 200.21
MDL Number MFCD00008508
Synonym Diphenylmethylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2715903

 
alfa aesar™
H2715903
1g Each for $36.03

AAH2715918

 
alfa aesar™
H2715918
50g Each for $527.97
Description & Specifications

Specifications

Formula Weight 200.21
Chemical Name or Material Methyldiphenylphosphine
Assay 99%
Molecular Formula (C6H5)2PCH3
MDL Number MFCD00008508
CAS 1486-28-8
Synonym Diphenylmethylphosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Boiling Point 160°C (15mmHg)
Quantity 1g

Applications
Methyldiphenylphosphine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.