missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyldiethoxysilane 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC392261000

 View more versions of this product

Catalog No. AC392261000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyldiethoxysilane
Clear colorless
2031-62-1
Glass bottle
Liquid
97%
(CH3CH2O)2SI(H)CH3
MFCD00026749
0.829g/mL
100g
Authentic
1.3740 to 1.3760 (20°C, 589nm)
≥96.0%
C5H14O2Si
134.25

Chemical Identifiers

97%
(CH3CH2O)2SI(H)CH3
MFCD00026749
≥96.0%
C5H14O2Si
134.25
Methyldiethoxysilane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger