Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyldichlorophosphine, 97+%

Chemical Identifiers

Chemical Name or Material Methyldichlorophosphine
CAS 676-83-5
Linear Formula (CH3)PCl2
Formula Weight 116.92
MDL Number MFCD00013616
UN Number UN2845
Synonym Methylphosphonous dichloride; Dichloromethylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3021103

  SDS
alfa aesar™
3021103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3021106

  SDS
alfa aesar™
3021106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.92
Odor Strong
TSCA TSCA
Quantity 1g
Color Colorless
Density 1.309g/mL
MDL Number MFCD00013616
UN Number UN2845
Chemical Name or Material Methyldichlorophosphine
Linear Formula (CH3)PCl2
CAS 676-83-5
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 81° to 82°C (729mmHg)
Flash Point 48° (120°F)
Health Hazard 2 GHS H Statement
H250-H226-H252-H314-H318
Catches fire spontaneously if exposed to air.
Flammable liquid and vapor.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay ≥98%
Refractive Index 1.4960
Synonym Methylphosphonous dichloride; Dichloromethylphosphine
Sensitivity Air & Moisture Sensitive
Physical Form Liquid
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H250-H226-H252-H314-H318
Catches fire spontaneously if exposed to air.
Flammable liquid and vapor.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.