missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylcyclopentane, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 345240250

Catalog No. AC345240250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methylcyclopentane
96-37-7
71.0°C
Authentic
Metal Drum
1.4077 to 1.4097
MFCD00001382
05, 27
cyclopentane, methyl, methyl cyclopentane, methylpentamethylene, unii-5g26cc1ask, methyl-cyclopentane, ccris 6058, hsdb 876, 5g26cc1ask, un2298 flammable liquid, 3-methylcyclopentyl
GDOPTJXRTPNYNR-UHFFFAOYSA-N
methylcyclopentane
7296
84.16
95%
0.7480g/mL
−4°C
94% min. (GC)
C6H12
C5H9CH3
25L
0.748
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,diethyl ether,ethanol,petroleum ether
CC1CCCC1
84.16
0.505 mPa.s (20°C)
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty

GHS Signal Word: Danger

EINECSNumber : 202-503-2

RTECSNumber : GY4640000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only