missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylcyclopentadienylmanganese tricarbonyl, 97%

Manufacturer:  Alfa Aesar™ 4712503

 View more versions of this product

Catalog No. AA4712503



Specifications

Specifications

Methylcyclopentadienylmanganese tricarbonyl
96°C (205°F)
Air Sensitive
Yellow
1g
C9H7MnO3
UN3281
MMT
218.09
97%
1.388g/mL
1.5873
233°C
1.5°C
12108-13-3
MFCD00001426
14,6223
Insoluble in water; Soluble in hydrocarbon solvents, alcohols, ether, acetone
Liquid

Chemical Identifiers

12108-13-3
C9H7MnO3
MFCD00001426
MMT
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.

RTECSNumber : OP1450000

TSCA : TSCA

SDS Documents