Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylcyclopentadienylmanganese tricarbonyl, 97%

Chemical Identifiers

Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl
CAS 12108-13-3
Molecular Formula C9H7MnO3
Formula Weight 218.09
MDL Number MFCD00001426
UN Number UN3281
Merck Index 14,6223
Synonym MMT
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4712503

  SDS
alfa aesar™
4712503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4712506

  SDS
alfa aesar™
4712506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4712514

  SDS
alfa aesar™
4712514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4712522

  SDS
alfa aesar™
4712522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.09
Chemical Name or Material Methylcyclopentadienylmanganese tricarbonyl
Density 1.388g/mL
TSCA TSCA
Refractive Index 1.5873
Physical Form Liquid
Assay 97%
Molecular Formula C9H7MnO3
MDL Number MFCD00001426
CAS 12108-13-3
UN Number UN3281
Merck Index 14,6223
Synonym MMT
Solubility Information Insoluble in water; Soluble in hydrocarbon solvents, alcohols, ether, acetone
Color Yellow
Health Hazard 1 Danger
Melting Point 1.5°C
Boiling Point 233°C
Quantity 1g
Flash Point 96°C (205°F)
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
RTECS Number OP1450000
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.