Learn More
Methylcyclopentadiene dimer, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 156850025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methylcyclopentadiene dimer | |
0.9400g/mL | |
26°C | |
95% | |
C12H16 | |
MFCD00082297 | |
0.94 | |
160.26 | |
95% |
26472-00-4 | |
200.0°C | |
Authentic | |
Glass bottle | |
1.4975 to 1.4995 | |
2.5kg | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene and ether | |
160.26 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 247-724-5
RUO â Research Use Only