missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylcyclohexane, 99+%

Manufacturer:  Alfa Aesar™ 3186122

 View more versions of this product

Catalog No. AA3186122



Specifications

Specifications

Methylcyclohexane
−4° (−25°F)
101°C
-126°C
108-87-2
MFCD00001497
505972
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
CC1CCCCC1
98.189
98.19
≥99%
0.771g/mL
1.4230
Colorless
100g
C7H14
UN2296
14,6047
UAEPNZWRGJTJPN-UHFFFAOYSA-N
methylcyclohexane
7962
Liquid

Chemical Identifiers

108-87-2
98.189
UAEPNZWRGJTJPN-UHFFFAOYSA-N
7962
CC1CCCCC1
C7H14
MFCD00001497
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
methylcyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P273-P243-P301+P330+P331-P403
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid release to the environment.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLCYCLOHEXANE

RTECSNumber : GV6125000

TSCA : TSCA

SDS Documents