Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylated Substance P, Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67835LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67835LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Regulatory Status RUO
Purification Method HPLC
Storage Requirements -30° to -10°C
Product Type Methylated Substance P, Certified Mass Spec Grade
Sequence RPKPQQFFGL-M(OMe)
Molecular Weight 1362.60
Format Lyophilized powder
Quantity 1 vial, 500 pmol