Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylated Substance P, Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67835ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67835ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 3 vials, 500 pmol/vial
Molecular Weight 1362.60
Sequence RPKPQQFFGL-M(OMe)
Format Lyophilized powder
Product Type Methylated Substance P, Certified Mass Spec Grade
Purification Method HPLC
Regulatory Status RUO
Storage Requirements -30° to -10°C