missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylaminoacetonitrile, TCI America™

Manufacturer:  TCI America M014025ML

 View more versions of this product

Catalog No. M014025ML



Specifications

Specifications

Methylaminoacetonitrile
Yellow
5616-32-0
MFCD00058964
PVVRRUUMHFWFQV-UHFFFAOYSA-N
2-(methylamino)acetonitrile
36652
Clear Liquid at 20°C
65°C
25mL
C3H6N2
1993
CNCC#N
70.095
70.10
≥98.0% (GC,T)

Chemical Identifiers

5616-32-0
70.095
PVVRRUUMHFWFQV-UHFFFAOYSA-N
2-(methylamino)acetonitrile
C3H6N2
MFCD00058964
36652
CNCC#N
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1465

TSCA : Yes

Recommended Storage : Freezer

Documents