missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Methylaminoacetonitrile hydrochloride, 99%
Manufacturer: Alfa Aesar™ H5521522
Specifications
Methylaminoacetonitrile hydrochloride | |
100g | |
C3H7ClN2 | |
MFCD00012587 | |
2-methylamino acetonitrile hydrochloride, acetonitrile, 2-methylamino-, hydrochloride 1:1, acetonitrile, methylamino-, monohydrochloride, cyanomethyl methyl ammonium chloride, methylamino acetonitrilhydrochlorid, methylaminoacetonitrile hydrochloride, methylaminoacetonitrile, hydrochloride, methylaminoacetonitrilehydrochloride, n-methylaminoacetonitrile hydrochloride, sarcosinenitrile hydrochloride | |
CNCC#N.Cl | |
106.553 | |
106.55 |
103°C to 106°C | |
25808-30-4 | |
CH3NHCH2CN·HCl | |
3908975 | |
DFKBQHBEROHUNF-UHFFFAOYSA-N | |
2-(methylamino)acetonitrile;hydrochloride | |
117641 | |
99% |
Chemical Identifiers
25808-30-4 | |
106.553 | |
DFKBQHBEROHUNF-UHFFFAOYSA-N | |
117641 | |
CNCC#N.Cl |
C3H7ClN2 | |
MFCD00012587 | |
2-methylamino acetonitrile hydrochloride, acetonitrile, 2-methylamino-, hydrochloride 1:1, acetonitrile, methylamino-, monohydrochloride, cyanomethyl methyl ammonium chloride, methylamino acetonitrilhydrochlorid, methylaminoacetonitrile hydrochloride, methylaminoacetonitrile, hydrochloride, methylaminoacetonitrilehydrochloride, n-methylaminoacetonitrile hydrochloride, sarcosinenitrile hydrochloride | |
2-(methylamino)acetonitrile;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 247-277-6
TSCA : Yes
Recommended Storage : Ambient temperatures