Please login to your online account to display your discounted pricing

Methyl Violet, Reagent Grade, LabChem™

$50.83 - $50.83

Chemical Identifiers

Chemical Name or Material Methyl Violet
CAS 8004-87-3
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C24H28N3Cl
Formula Weight 393.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC171907

 
lab chem
LC171907
Amber Glass 10g Each for $50.83
Description & Specifications

Specifications

Formula Weight 393.96
Chemical Name or Material Methyl Violet
Quantity 10g
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula C24H28N3Cl
CAS 8004-87-3
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide
Melting Point 137°C
Color Greenish
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.

Room Temperature