Please login to your online account to display your discounted pricing

Methyl Violet, 0.02% Aqueous, pH 0.0 to 1.4 Green to Blue, Certified, LabChem™

$75.26 - $75.26

Chemical Identifiers

Chemical Name or Material Methyl Violet
CAS 7732-18-5
CAS Max % 99.98
CAS 8004-87-3
CAS Max % 0.02
Name Note 0.02% Aqueous, pH 0.0 to 1.4 Green to Blue
Purity Grade Certified Grade
Linear Formula C24H28N3Cl
Formula Weight 393.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC172001

 
lab chem
LC172001
Poly Bottle 500mL Each for $75.26
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 393.96
Chemical Name or Material Methyl Violet
Quantity 500mL
Name Note 0.02% Aqueous, pH 0.0 to 1.4 Green to Blue
Physical Form Liquid
Linear Formula C24H28N3Cl
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.98
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 8004-87-3
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.02
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Dark purple
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature