Please login to your online account to display your discounted pricing

Methyl vinyl sulfone, 98%, stabilized, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC306730010

 View more versions of this product

Catalog No. AC306730010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 106.14
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.21g/mL
CAS 650166
Chemical Name or Material Methyl vinyl sulfone
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note Stabilized
Assay Percent Range 98%
Molecular Formula C3H6O2S
Linear Formula HC=CHSOCH3
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Stabilizer 150ppm 4-tert.-butylpenol
MDL Number MFCD00007567
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Fatal in contact with skin.
Causes serious eye damage.

Specifications

Chemical Name or Material Methyl vinyl sulfone
Name Note Stabilized
CAS 650166
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H6O2S
Linear Formula HC=CHSOCH3
Formula Weight 106.14
MDL Number MFCD00007567
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Fatal in contact with skin.
Causes serious eye damage.

SDS & Safety Documents