Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol

Chemical Identifiers

Chemical Name or Material Methyl vinyl sulfone
CAS 3680-02-2
Name Note stabilized with 200ppm 4-tert-butylphenol
Molecular Formula C3H6O2S
Formula Weight 106.15
MDL Number MFCD00007567
UN Number UN2810
Beilstein 878198
Synonym (Methylsulfonyl)ethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0151903

  SDS
alfa aesar™
L0151903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0151906

  SDS
alfa aesar™
L0151906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.15
Chemical Name or Material Methyl vinyl sulfone
Assay 95%
MDL Number MFCD00007567
CAS 3680-02-2
UN Number UN2810
Health Hazard 2 GHS H Statement
H310-H318-H302
Fatal in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Name Note stabilized with 200ppm 4-tert-butylphenol
Molecular Formula C3H6O2S
Beilstein 878198
Synonym (Methylsulfonyl)ethylene
Health Hazard 1 Danger
Boiling Point 115° to 117°C (19mmHg)
Quantity 1g
Flash Point >109°C (228°F)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number WR6825000
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H310-H318-H302
Fatal in contact with skin.
Causes serious eye damage.
Harmful if swallowed.

2° to 8°C