missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl vinyl ketone, tech. 90%, stab., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11910AE

 View more versions of this product

Catalog No. AAA11910AE



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl Vinyl Ketone
78-94-4
0.856
-6°C (21°F)
C4H6O
MFCD00008777
UN1251
14,6134
FUSUHKVFWTUUBE-UHFFFAOYSA-N
but-3-en-2-one
6570
70.09
Technical
Stabilized
°C to 7°C
32°C to 34°C (60 mm)
90%, Stabilized with 0.5% Hydroquinone
1.412
100 mL
506021
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
CC(=O)C=C
70.09
CHEBI:48058
90%
Liquid

Chemical Identifiers

78-94-4
70.09
FUSUHKVFWTUUBE-UHFFFAOYSA-N
6570
but-3-en-2-one
C4H6O
MFCD00008777
methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
CHEBI:48058
CC(=O)C=C
Safety and Handling

Safety and Handling

GHS H Statement
H224-H300-H330-H314-H318

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H300+H310+H330-H314-H317-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL VINYL KETONE, STABILIZED

EINECSNumber : 201-160-6

RTECSNumber : EM9800000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only