missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl vinyl ketone, tech. 90%, stab.

Manufacturer:  Alfa Aesar™ A11910AC

 View more versions of this product

Catalog No. AAA11910AC


 

Specifications

Specifications

Methyl vinyl ketone
32°C to 34°C (60mmHg)
25mL
78-94-4
90%
C4H6O
UN1251
14,6134
FUSUHKVFWTUUBE-UHFFFAOYSA-N
but-3-en-2-one
6570
70.09
−6°C (21°F)
-7°C
Stabilized
Technical Grade
90%, Stabilized with 0.5% Hydroquinone
MFCD00008777
506021
3-Buten-2-one
CC(=O)C=C
70.091
CHEBI:48058
Liquid

Chemical Identifiers

78-94-4
70.091
FUSUHKVFWTUUBE-UHFFFAOYSA-N
6570
C4H6O
MFCD00008777
3-Buten-2-one
CHEBI:48058
Safety and Handling

Safety and Handling

GHS H Statement
H224-H300-H330-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL VINYL KETONE, STABILIZED

RTECSNumber : EM9800000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents