Please login to your online account to display your discounted pricing

Methyl Vinyl Ketone, Stabilized ca 95%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128000010

 View more versions of this product

Catalog No. AC128000010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 70.09
Quantity 1L
Color Clear colorless to pale yellow, may discolor to brown on storage
Water ∽3%
Density 0.8640g/mL
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
CAS 123-31-9
CAS 7732-18-5
Packaging Glass bottle
CAS 78-94-4
CAS 75-05-8
Molecular Formula C4H6O
CAS 64-19-7
Infrared Spectrum Authentic
Synonym 3-Buten-2-one
Chemical Name or Material Methyl vinyl ketone
Linear Formula H2C=CHCOCH3
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Assay Percent Range ca. 95%
Health Hazard 1 Danger

Specifications

Chemical Name or Material Methyl vinyl ketone
CAS 78-94-4
CAS 75-05-8
CAS 64-19-7
CAS 123-31-9
CAS 7732-18-5
Assay ≥94%
Assay Percent Range ca. 95%
Molecular Formula C4H6O
Linear Formula H2C=CHCOCH3
Formula Weight 70.09
Synonym 3-Buten-2-one
Accessories

Danger

SDS & Safety Documents