Please login to your online account to display your discounted pricing

Methyl Vinyl Ketone, Stabilized ca 95%, ACROS Organics™

$60.72 - $800.42

Chemical Identifiers

Chemical Name or Material Methyl Vinyl Ketone
CAS 78-94-4
CAS 75-05-8
CAS 64-19-7
CAS 123-31-9
CAS 7732-18-5
Assay ≥94%
Assay Percent Range ca. 95%
Molecular Formula C4H6O
Linear Formula H2C=CHCOCH3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC128000010

  SDS
thermo fisher scientific
128000010
Glass bottle 1L Each for $383.32

AC128000050

  SDS
thermo fisher scientific
128000050
Glass bottle 5mL Each for $60.72

AC128000025

  SDS
thermo fisher scientific
AC128000025
Glass Bottle 2.5L Each for $800.42
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94%
Formula Weight 70.09
Quantity 1L
Color Clear colorless to pale yellow, may discolor to brown on storage
Water ∽3%
Density 0.8640g/mL
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
CAS 123-31-9
CAS 7732-18-5
Chemical Name or Material Methyl vinyl ketone
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H6O
CAS 78-94-4
Synonym 3-Buten-2-one
CAS 75-05-8
CAS 64-19-7
Assay Percent Range ca. 95%
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Linear Formula H2C=CHCOCH3
Health Hazard 1 Danger
Safety and Handling

Danger