Please login to your online account to display your discounted pricing

Methyl Vinyl Ketone, Stabilized ca 95%, ACROS Organics™


Quantity 2.5L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC128000025

 View more versions of this product

Catalog No. AC128000025

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Methyl Vinyl Ketone
Flash Point −7°C
Boiling Point 80.0°C
Color Clear colorless to pale yellow, may discolor to brown on storage
Quantity 2.5L
CAS 78-94-4
Health Hazard 1 Danger
CAS 75-05-8
Density 0.8640g/mL
CAS 64-19-7
CAS 123-31-9
Infrared Spectrum Authentic
CAS 7732-18-5
Packaging Glass Bottle
Physical Form Liquid
Assay ≥94%
Assay Percent Range ca. 95%
Stabilizer 0.7% acetonitrile max., 0.4 to 1.0% acetic acid, approx. 0.3% hydroquinone
Molecular Formula C4H6O
Water ∽3%
Linear Formula H2C=CHCOCH3
Formula Weight 70.09
Fieser 01,697; 02,283; 05,464; 06,407; 07,247; 10,272; 12,329; 13,193
Synonym 3-Buten-2-one

Specifications

Chemical Name or Material Methyl Vinyl Ketone
CAS 78-94-4
CAS 75-05-8
CAS 64-19-7
CAS 123-31-9
CAS 7732-18-5
Assay ≥94%
Assay Percent Range ca. 95%
Molecular Formula C4H6O
Linear Formula H2C=CHCOCH3
Formula Weight 70.09
Synonym 3-Buten-2-one
Accessories

Danger

SDS & Safety Documents