missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Vanillate 98.0+%, TCI America™
Supplier: TCI America M173825G
Specifications
| Methyl Vanillate | |
| 65°C | |
| C9H10O4 | |
| 25 g | |
| BVWTXUYLKBHMOX-UHFFFAOYSA-N | |
| methyl 4-hydroxy-3-methoxybenzoate | |
| 19844 | |
| 182.18 | |
| Crystalline Powder |
| 3943-74-6 | |
| 287°C | |
| MFCD00008438 | |
| methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx | |
| COC1=C(C=CC(=C1)C(=O)OC)O | |
| 182.175 | |
| CHEBI:46477 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3943-74-6 | |
| 182.175 | |
| BVWTXUYLKBHMOX-UHFFFAOYSA-N | |
| 19844 | |
| methyl 4-hydroxy-3-methoxybenzoate |
| C9H10O4 | |
| MFCD00008438 | |
| methyl vanillate, methyl 3-methoxy-4-hydroxybenzoate, vanillic acid, methyl ester, 4-hydroxy-3-methoxybenzoic acid methyl ester, methylvanillate, benzoic acid, 4-hydroxy-3-methoxy-, methyl ester, vanillic acid methyl ester, unii-2hxg8qso3d, 2hxg8qso3d, vxx | |
| CHEBI:46477 | |
| COC1=C(C=CC(=C1)C(=O)OC)O |
Safety and Handling
RTECSNumber : DH2430000
TSCA : Yes