missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl valerate, 99%, Thermo Scientific Chemicals
$184.03 - $184.03
Chemical Identifiers
CAS | 624-24-8 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009478 |
InChI Key | HNBDRPTVWVGKBR-UHFFFAOYSA-N |
Synonym | methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn |
PubChem CID | 12206 |
IUPAC Name | methyl pentanoate |
SMILES | CCCCC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2482836
|
Thermo Scientific Chemicals
B2482836 |
500 g |
Each for $184.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
624-24-8 | |
116.16 | |
HNBDRPTVWVGKBR-UHFFFAOYSA-N | |
12206 | |
CCCCC(=O)OC |
C6H12O2 | |
MFCD00009478 | |
methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn | |
methyl pentanoate |
Specifications
624-24-8 | |
0.885 | |
22°C (71°F) | |
1.397 | |
500 g | |
1741905 | |
HNBDRPTVWVGKBR-UHFFFAOYSA-N | |
methyl pentanoate | |
12206 | |
99% |
-91°C | |
126°C to 128°C | |
C6H12O2 | |
MFCD00009478 | |
UN3272 | |
methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn | |
CCCCC(=O)OC | |
116.16 | |
116.16 | |
Methyl valerate |
Safety and Handling
GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 210-838-0
RTECSNumber : YV7750500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only