missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (Triphenylphosphoranylidene)acetate, TCI America™

Manufacturer:  TCI America T1363100G

 View more versions of this product

Catalog No. T1363100G



Specifications

Specifications

Methyl (Triphenylphosphoranylidene)acetate
169°C
2605-67-6
MFCD00008455
NTNUDYROPUKXNA-UHFFFAOYSA-N
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
17453
Crystal-Powder at 20°C
White-Yellow
100g
C21H19O2P
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, carbomethoxymethylene triphenylphosphorane, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
COC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
334.355
334.35
≥98.0% (HPLC,T)

Chemical Identifiers

2605-67-6
334.355
NTNUDYROPUKXNA-UHFFFAOYSA-N
17453
COC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
C21H19O2P
MFCD00008455
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, carbomethoxymethylene triphenylphosphorane, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents