missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (triphenylphosphoranylidene)acetate, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC169565000

 View more versions of this product

Catalog No. AC169565000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

for C-C Bond Formation (Olefination)
Specifications

Specifications

Methyl (triphenylphosphoranylidene)acetate
Glass bottle
164.0°C to 169.0°C
2605-67-6
C21H19O2P
01,112; 01,1081; 08,456; 09,426; 10,363; 13,277
NTNUDYROPUKXNA-UHFFFAOYSA-N
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
17453
Powder
Authentic
White
500g
98%
(C6H5)3P=CHCO2CH3
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, carbomethoxymethylene triphenylphosphorane, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
COC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
334.355
334.34
98%

Chemical Identifiers

2605-67-6
334.355
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, carbomethoxymethylene triphenylphosphorane, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
C21H19O2P
NTNUDYROPUKXNA-UHFFFAOYSA-N
17453
COC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
SDS Documents